(1S,3R)-(-)-Camphoric acid ≥99%

Supplier: 0000027161

Synonyms: (1S,3R)-1,2,2-Trimethylcyclopentane-1,3-dicarboxylic acid

CDX-C0144-G001 CDX-C0144-G010
102989-496EA 166.15 USD
102989-496 102989-498
(1S,3R)-(-)-Camphoric acid ≥99%
(1S,3R)-(-)-Camphoric acid

Chirality inducing agent. Useful tool as resolving agent to separate the isomers and purify racemic mixtures (e.g. DL-lysine) by fractionally crystallizing the product to obtain the pure isomer (e.g. D-lysine). Can also be used as building block.

Formula: C₁₀H₁₆O₄
Storage Temperature: Freezer
MDL Number: MFCD00064937
CAS Number: 560-09-8

Order Now

Specification Test Results

Source/Host Synthetic
Purity 99% (T)
Appearance/Color White crystalline.
Solubility Soluble in ethanol, methanol or water.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR