Guanidinium hydrochloride, Bioreagent

Supplier: Avantor Performance Materials
Warning

Synonyms: Guanidinium chlorideGuanidine hydrochlorideGuanidine chlorideAminoformamidine hydrochlorideGuanadine hydrochlorideAminomethanamidine hydrochlorideGuanidine HCI

VWRVVWRB287-12KG VWRVVWRB287-5KG VWRVVWRB287-50KG
VWRB287-5KGEA 2189.43 USD
VWRB287-5KG VWRB287-50KG VWRB287-12KG
Guanidinium hydrochloride, Bioreagent
Guanidinium hydrochloride

Material is of non-animal, non-human origin.


  • Bioprocessing
  • Bioreagent

Formula: NH₂C(=NH)NH₂·HCl
MW: 95.53 g/mol
Melting Pt: 181…187 °C
Density: 1.354 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013026
CAS Number: 50-01-1
https://us-prod2.vwr.com/stibo/web/std.lang.all/74/24/2297424.jpg

Order Now

Specification Test Results

Abs. 230nm (6M, Water) ≤ 0.2
Abs. 260nm (6M, Water) ≤ 0.03
Abs. 275nm (6M, Water) ≤ 0.03
Arsenic ≤ 0.0500 ppm
Assay 99.5-101.0%
Copper ≤ 0.200 ppm
Cyanide ≤ 0.1 ppm
Dnase None
Iron ≤ 2.0 ppm
Lead ≤ 0.500 ppm
Loss on Drying ≤ 0.5%
Nitrate ≤ 0.005%
Protease None
Residue on Ignition ≤ 0.05%
Solubility (6M, Water) Pass
Sulfate ≤ 0.005%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR