Zinc salicylate trihydrate ≥95.0%

Supplier: Spectrum Chemical Mfg. Corp.

Z1042-100GM Z1042-500GM
700016-296EA 104.02 USD
700016-296 700016-298
Zinc salicylate trihydrate ≥95.0%
Zinc salicylate trihydrate
Storage Temperature: Ambient
CAS Number: 16283-36-6

Order Now

Specification Test Results

Assay 95.0 - 103.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR