DL-α-Tocopherol (Vitamin E), 1.1 IU/mg ≥96.0% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: (±)-α-Tocopherol, Vitamin E Alcohol, all-rac-α-Tocopherol, 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-benzopyran-6-ol, 2,5,7,8-Tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-ol, DL-alpha-Tocopherol, Vitamin E

VI135-100ML VI135-1LTGL VI135-20KGBL
89085-250EA 219.58 USD
89085-250 89085-252 75812-602
DL-α-Tocopherol (Vitamin E), 1.1 IU/mg ≥96.0% USP
DL-α-Tocopherol

Vitamin E, 1 mg = 0.74 alpha-tocopherol equivalents, USP. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₂₉H₅₀O₂
MW: 430.71 g/mol
Density: 0.950
Storage Temperature: Refrigerator
MDL Number: MFCD00072051
CAS Number: 10191-41-0

Order Now

Specification Test Results

Assay (C19H50O₂) 96.0 - 102.0 %
Acidity To Pass Test
Identification To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR