Trimellitic acid tris(2-ethylhexyl) ester ≥99%
Supplier: Spectrum Chemical Mfg. Corp.
Tri(2-ethylhexyl) Trimellitate is used as a plasticizer for cosmetic and general use.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₃₃H₅₄O₆ Density: 0.99 Storage Temperature: Ambient |
MDL Number:
MFCD00053799 CAS Number: 3319-31-1 |
Specification Test Results
Assay | Approx. 99 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...