Formaldehyde sodium bisulfite ≥93%
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Sodium Hydroxymethanesulphonate, Sodium Hydroxymethanesulfonate, Hydroxymethanesulfonic acid sodium salt, Hydroxymethanesulphonic acid sodium salt
Sodium Formaldehyde Bisulfite is used in the preparation of oil-soluble sulfonate additives used in improving anticorrosive, dispersant and antioxygenic properties of lubricant oils.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
CH₄O₄S MW: 134.08 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00044664 CAS Number: 870-72-4 |
Specification Test Results
Assay (Anhydrous Basis) | 93 % |
Moisture (KF) | max. 6.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...