N-Succinimidyl Acrylate ≥98%
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Synonyms:
N-Acryloxysuccinimide, 2,5-Dioxopyrrolidin-1-yl acrylate, N-Acryloyloxysuccinimide, Acrylic Acid N-Succinimidyl Ester
S1722-5GM
S1722-25GM
10860-560EA
177.05
USD
10860-560
10860-562
N-Succinimidyl Acrylate ≥98%
N-Succinimidyl Acrylate
Storage temperature - FREEZE: Keep tightly closed.
GHS hazard : Exclamation mark
Caution: Storage temperature - FREEZE: Keep tightly closed.
Formula:
C₇H₇NO₄ MW: 169.14 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00078261 CAS Number: 38862-24-7 |
Specification Test Results
Purity (GC) | 98 % |
Melting Point | 70.0°-72.0°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...