Nigrosine, water soluble

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Acid Black 2

NI115-100GM NI115-25KGBL NI115-25GM
700010-112EA 179.1 USD
700010-112 700010-116 700010-114
Nigrosine, water soluble
Nigrosine

Nigrosin, Water Soluble, a mixture of synthetic black dyes, is used in biology for negative staining of bacteria.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Ambient
MDL Number: MFCD00044681
CAS Number: 8005-03-6

Order Now

Specification Test Results

Absorption Max Approx. 570 NM

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR