Cellulose, microcrystalline NF
Supplier: SPECTRUM CHEMICAL MFG CORP
C1679-100GM
C1679-2.5KG
C1679-50KGBL
C1679-500GM
C1679-12KGBL
95034-600EA
180.68
USD
95034-600
95034-602
700013-062
95034-604
75810-470
Cellulose, microcrystalline NF
Cellulose
Microcrystalline Cellulose, NF is an excipient in pharmaceutical formulations acting as a binder in tablet formation. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
(C₁₂H₂₂O₁₁)n Storage Temperature: Ambient |
MDL Number:
MFCD00081512 CAS Number: 9004-34-6 |
Specification Test Results
pH of Solution | 5.0 - 7.5 |
Conductivity | max. 75 Us/Cm |
Loss On Drying | max. 7.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...