meso-Erythritol

Supplier: Spectrum Chemical Mfg. Corp.

ER123-5GM ER123-25GM
700003-032EA 57 USD
700003-032 700003-030
meso-Erythritol
meso-Erythritol

meso-Erythritol is a four-carbon sugar that is found in algae, fungi, and lichens. It is twice as sweet as sucrose and can be used as a coronary vasodilator. It is used in cosmetic and personal care products as a humectant and skin conditioning agent with moisturizing properties. It is used in skin care, oral care, cleansers, and toners. It is easily soluble in cold water

Formula: C₄H₁₀O₄
Storage Temperature: Ambient
MDL Number: MFCD00004710
CAS Number: 149-32-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR