Maltitol ≥95%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: (2S,3R,4R,5R)-4-(((2R,3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexane-1,2,3,5,6-pentaol

M3108-25GM M3108-100GM
700009-678EA 145.9 USD
700009-678 700009-676
Maltitol ≥95%
Maltitol

Maltitol, also known as Amalty, is a sugar alcohol and used as a sugar substitute. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Formula: C₁₂H₂₄O₁₁
Storage Temperature: Ambient
MDL Number: MFCD00006600
CAS Number: 585-88-6

Order Now

Specification Test Results

Assay (By TLC) Approx. 95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR