Hydroxyethyl cellulose, ~3400 cP (1%, H₂O)

Supplier: Spectrum Chemical Mfg. Corp.

H1148-12KG H1148-100GM H1148-45KGBL H1148-500GM H1148-2.5KG
700013-000EA 3378.25 USD
700013-000 700012-998 700013-004 700013-006 700013-002
Hydroxyethyl cellulose, ~3400 cP (1%, H₂O)
Hydroxyethyl cellulose

Hydroxyethyl Cellulose, 3,400 cps is a thickening and gelling agent created from cellulose, most commonly used in cleaning solutions, cosmetics and other products used in the household. It usually comes in capsule form.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Ambient
MDL Number: MFCD00072770
CAS Number: 9004-62-0

Order Now

Specification Test Results

Viscosity of a 1% Aqueous Solution Approx. 3,400 cP
Loss On Drying @ 105 C max.5.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR