Geranium oil Chinese Type

Supplier: Spectrum Chemical Mfg. Corp.

G1198-100ML G1198-500ML
700010-646EA 175.44 USD
700010-646 700010-648
Geranium oil Chinese Type
Geranium oil

Geranium Oil, Chinese Type, also called Pelargonium graveolens oil or rose geranium oil, is an essential oil typically used in massage and aromatherapy applications.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Ambient
MDL Number: MFCD00240716
CAS Number: 8000-46-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR