Oleic acid cholesteryl ester

Supplier: Spectrum Chemical Mfg. Corp.

C3824-1GM C3824-250MG C3824-500MG
700005-434EA 261.59 USD
700005-434 700005-436 700005-438
Oleic acid cholesteryl ester
Oleic acid cholesteryl ester

Cholesteryl Oleate is an esterified form cholesterol found in plasma.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₄₅H₇₈O₂
Storage Temperature: Freezer
MDL Number: MFCD00003645
CAS Number: 303-43-5

Order Now

Specification Test Results

Melting Point 42 - 48 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR