Chlorophyllin, coppered trisodium salt

Supplier: Spectrum Chemical Mfg. Corp.

C3999-25GM C3999-100GM C3999-5GM C3999-50KGBL
700008-926EA 124.2 USD
700008-926 700008-924 700008-930 700008-928
Chlorophyllin, coppered trisodium salt
Chlorophyllin, coppered trisodium salt

Chlorophyllin Sodium Copper Salt | Non-graded Chlorophyllin Sodium Copper Salt supplied by Spectrum is indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₃₄H₃₁CuN₄Na₃O₆
Storage Temperature: Ambient
MDL Number: MFCD00012149
CAS Number: 11006-34-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR