Chitin

Supplier: SPECTRUM CHEMICAL MFG CORP

C2020-100GM C2020-25GM
700012-934EA 159.4 USD
700012-934 700012-936
Chitin
Chitin

Chitin is used as a binder in fabrics, dyes, and adhesives.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₃₀H₅₀N₄O₁₉
MW: 233.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00466914
CAS Number: 1398-61-4

Order Now

Specification Test Results

Ash Content max. 5 %
Loss On Drying max. 10 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR