Calamine ≥98.0%, powder USP

Supplier: SPECTRUM CHEMICAL MFG CORP

CA115-2.5KG CA115-500GM CA115-12KG CA115-125GM
95032-194EA 551.25 USD
95032-194 95032-196 700013-948 95032-192
Calamine ≥98.0%, powder USP
Calamine

CALAMINE, POWDER, USP is a zinc based active ingredient use in topical medications to relieve pain and itching to treat conditions such as sunburn, rashes, poison ivy, poison oak, chickenpox, and insect bites and stings and an astringent to dry weeping or oozing blisters and acne abscesses. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Storage Temperature: Ambient
CAS Number: 8011-96-9

Order Now

Specification Test Results

Assay (ZnO) 98.0 - 100.5 %
Microbial Limits To Pass Test
Loss On Ignition max. 2.0 %
Acid-Insoluble Substances max. 2.0 %
Alkaline Substances To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR