β-Carotene ≥96%, crystalline powder

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 2,2'-((1E,3E,5E,7E,9E,11E,13E,15E,17E)-3,7,12,16-Tetramethyloctadeca-1,3,5,7,9,11,13,15,17-nonaene-1,18-diyl)bis(1,3,3-trimethylcyclohex-1-ene), Provitamin A

CA200-25GM CA200-5GM
700010-256EA 316.1 USD
700010-256 700010-258
β-Carotene ≥96%, crystalline powder
beta-Carotene

Storage temperature - LIGHT SENSITIVE, AIR SENSITIVE, MOISTURE SENSITIVE: FREEZE: Keep tightly closed in light-resistant containers. Store under Nitrogen.

Formula: C₄₀H₅₆
MW: 536.89 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00001556
CAS Number: 7235-40-7

Order Now

Specification Test Results

Assay 96 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR