Arbutin ≥99.5%

700004-158EA 19747.2 USD
700004-158 700004-154 700004-156 700004-160 700004-162
Arbutin ≥99.5%
Arbutin

Arbutin is a derivative of hydroquinone used in skin-whitening ingredients. It is reported to inhibit the activity of tyrosinase, which is a key enzyme for melanin biosynthesis. This acts to effectively prevent the anomalous increase of melanin which is blamed for "age or sun spots and freckles.

Formula: C₁₂H₁₆O₇
Storage Temperature: Ambient
MDL Number: MFCD00016915
CAS Number: 497-76-7

Order Now

Specification Test Results

Assay 99.5 % min.
Loss On Drying max.0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR