4-Aminopyridine ≥99.0%, Reagent Grade

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Pyridin-4-amine, 4-Pyridylamine

AM164-100GM AM164-25GM
10841-738EA 297.55 USD
10841-738 10841-740
4-Aminopyridine ≥99.0%, Reagent Grade
4-Aminopyridine

4-Aminopyridine, Reagent is an organic compound and is a useful pharmacological tool in studying various potassium conductances in physiology and biophysics. It has also been utilized to manage symptoms of multiple sclerosis. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.


GHS hazard : Skull and crossbones;Exclamation mark;Health hazard UN2671

Formula: C₅H₆N₂
Storage Temperature: Ambient
MDL Number: MFCD00006439
CAS Number: 504-24-5
UN: 2671
ADR: 6.1,II

Order Now

Specification Test Results

Purity (GC) 99.0 % min.
Melting Point 157.0 - 162.0°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR