Hydrogen peroxide 3% (w/w) in water stabilized, Fluka™

Supplier: Honeywell Research Chemicals
Danger


Stabilization: Stabilised with acetanilide ~200 ppm

323381-25ML 323381-4L 323381-500ML
BJ323381-25MLEA 63.59 USD
BJ323381-25ML BJ323381-4L BJ323381-500ML
Hydrogen peroxide 3% (w/w) in water stabilized, Fluka™
Hydrogen peroxide

contains ∼200 ppm acetanilide as stabilizer

Formula: H₂O₂
Melting Pt: –26 °C
Density: 1.00 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011333
CAS Number: 7722-84-1
UN: 2014
ADR: 5.1,II

Order Now

Specification Test Results

Appearance (colorless liquid) complying
Assay 3.0 - 4.0 Gew.%
pH (direct, 20 °C) 4 - 7
Stabilizer (Acetanilide) 150 - 250 ppm
Contains ~200 ppm acetanilide as stabilizer


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR