Ammonia solution ∼25% (NH₃ basis) in water, Puriss. p.a. ISO, Reag. Ph. Eur., Fluka™
Supplier: Honeywell Research Chemicals
|
Danger
|
30501-5L
30501-4X5L
30501-6X1L
BJ30501-1LEA
102.84
USD
BJ30501-1L
BJ30501-1L-GL
BJ30501-2.5L
BJ30501-2.5LGL
BJ30501-5L
BJ30501-4X2.5L
BJ30501-4X5L
BJ30501-6X1L
BJ30501-6X1LGL
Ammonia solution ∼25% (NH₃ basis) in water, Puriss. p.a. ISO, Reag. Ph. Eur., Fluka™
Ammonia solution
Formula:
NH₃ Boiling Pt: 37.7 °C (1.013 hPa) Melting Pt: -92 °C Density: 0.90 g/cm³ (20 °C) Flash Pt: 30.2 %(V) Storage Temperature: Ambient |
MDL Number:
MFCD00066650 CAS Number: 1336-21-6 UN: 2672 ADR: 8,III |
Specification Test Results
Assay | Min. 25.0 % |
Density (D 20/20) | 0.892 - 0.910 |
Non-volatile matter | Max. 0.0018 % |
Silver (Ag) | Max. 0.02 ppm |
Aluminium (Al) | Max. 0.5 ppm |
Gold (Au) | Max. 0.1 ppm |
Barium (Ba) | Max. 0.05 ppm |
Bismuth (Bi) | Max. 0.1 ppm |
Calcium (Ca) | Max. 1 ppm |
Cadmium (Cd) | Max. 0.05 ppm |
Cobalt (Co) | Max. 0.05 ppm |
Chromium (Cr) | Max. 0.05 ppm |
Copper (Cu) | Max. 0.1 ppm |
Iron (Fe) | Max. 0.1 ppm |
Gallium (Ga) | Max. 0.02 ppm |
Indium (In) | Max. 0.02 ppm |
Potassium (K) | Max. 0.5 ppm |
Lithium (Li) | Max. 0.02 ppm |
Magnesium (Mg) | Max. 0.1 ppm |
Manganese (Mn) | Max. 0.05 ppm |
Molybdenum (Mo) | Max. 0.05 ppm |
Sodium (Na) | Max. 1 ppm |
Nickel (Ni) | Max. 0.05 ppm |
Lead (Pb) | Max. 0.05 ppm |
Platinum (Pt) | Max. 0.1 ppm |
Tin (Sn) | Max. 0.1 ppm |
Strontium (Sr) | Max. 0.2 ppm |
Titanium (Ti) | Max. 0.1 ppm |
Thallium (Tl) | Max. 0.05 ppm |
Zinc (Zn) | Max. 0.1 ppm |
Heavy metals (as Pb) | Max. 1 ppm |
Carbonate (CO₃) | Max. 60 ppm |
Carbonate (as CO₂) | Max. 10 ppm |
Chloride (Cl) | Max. 0.5 ppm |
Phosphate (PO₄) | Max. 0.5 ppm |
Silicate (as SiO₂) | Max. 10 ppm |
Sulfate (SO₄) | Max. 2 ppm |
Sulfide (S) | Max. 0.2 ppm |
KMnO₄ reducing matter (as O) | Max. 0.0005 % |
Readily oxidizable substances | complying |
Pyridine, related substances (as Pyridine) | Max. 2 ppm |
Pyridine | Max. 0.00005 % |
Appearance of the solution | complying |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...