CD3, T-cell IHC Positive Control Slides, Newcomer Supply

Supplier: Newcomer Supply
3770A 3770B
75877-130BX 91.27 USD
75877-130 75877-132
CD3, T-cell IHC Positive Control Slides, Newcomer Supply
Slides Microscopy Control Slides
CD3, T-cell control slides contain a section of positive staining tonsil.

  • Tissue type: (+) Tonsil
  • All control tissue is sectioned on Superfrost® Plus slides
  • Staining validation technical memo included in each set
  • All tissues are formalin fixed, paraffin embedded FFPE
  • All slides are air dried
  • IHC slides are cut at 4 µm
  • Standard tissue placement is mid-high on slide

Newcomer supply CD3, T-cell control slides are for the positive immunohistochemical staining of T-Cells

Staining Results:
T-cell positive expression will stain brown membrane staining.
Nuclei will stain blue.

The enclosed positive control slides are intended to be used to verify histological techniques and reagent reactivity. These slides are to be used for the qualitative purpose of determining positive or negative results, and are not intended to be used for any quantitative purpose.

CD3 is considered to be a pan-T-cell marker, and widely used in detection of T-cell malignancies, both immature and mature.

Ordering information: All controls are cut in serial sections.
10 set = 10 unstained slides and 1 stained slide.
98 set = 98 unstained slides and 2 stained slides. (First & Last Slides are Stained)
Reactivity of IHC control slides are guaranteed for one year from the date of receipt and should be revalidated after one year to verify continued reactivity.
Please note the following: Allow 2-3 weeks for delivery for large sets. In human tissue unless otherwise indicated.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR