Magnetic Plates, OZ Biosciences
Supplier: OZ Biosciences
MF10000 MF14000 MF10096
103258-352EA 683.31 USD
103258-352 103258-354 103549-570
Magnetic Plates, OZ Biosciences
Magnets
Magnetic plate for all cell culture supports applications.
  • Can be easily cleaned and decontaminated with 70% ethanol
  • Can be used at room temperature, 37°C, +4°C, etc.
  • Compatible with culture plates from most common suppliers
  • Solid, completely reusable
Suitable for all Magnetofection reagents and all cell culture dishes and flasks and can be used within incubators and with robots.