Economy Constant Temperature Water Bath, Yamato
Supplier: Yamato
BM-100 BM-302A BM-200 BM-500 BM-401
75846-736EA 1045.56 USD
75846-736 76443-162 75846-396 33995-007 75846-632
Economy Constant Temperature Water Bath, Yamato
Baths Water Baths, Unstirred
The compact design and simple set-up systems make these water baths easy to use with rotary evaporators.
  • Smaller baths equipped with a drain
  • Various sizes available
  • Bath protective cover
  • Thermal fuse
The larger sizes are equipped with digital setting by UP/DOWN keys.
All models feature thermal fuses for safety.