Di-p-tolylphosphine ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

15-1820-2G 15-1820-10G
100196-092EA 213.72 USD
100196-092 100196-090
Di-p-tolylphosphine ≥99%
Di-p-tolylphosphine

Phosphine

Formula: (CH₃C₆H₄)₂PH
MDL Number: MFCD01630843
CAS Number: 1017-60-3
UN: 2845
ADR: 4.2,I

Order Now

Specification Test Results

Assay 99%
Color Colorless
Form Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR