Acetic acid glacial ≥99.7% ACS
Supplier: AQUA SOLUTIONS MS
|
Danger
|
A1100-500ML
A1100-4L
A1100-20L
A1100-2.5L
101204-870EA
78.12
USD
101204-870
101203-216
101204-078
101203-792
Acetic acid glacial ≥99.7% ACS
Acetic acid
Formula:
H₃CCOOH |
MDL Number:
MFCD00036152 CAS Number: 64-19-7 UN: 2789 ADR: 8,II |
|
Specification Test Results
Assay | ≥ 99.7% CH3COOH |
Residue after evaporation | Max. 0.001% |
Acetic anhydride [(CH3CO)20] | Max. 0.01% |
Color (APHA) | Max. 10 |
Chloride (Cl) | Max. 1 ppm |
Heavy Metals, as Pb | Max. 0.5 ppm |
Iron | Max. 0.2 ppm |
Sulfate (SO4) | Max. 1 ppm |
Titrable Base | Max. 0.0004 meq |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...