Sodium thiosulfate 0.05 M Na2S2O3 (0.05N) volumetric solution, Fluka™
Supplier: Honeywell Research Chemicals
Synonyms:
di-Sodium thiosulfate, Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate, Sodiumthiosulfate, Sodium hyposulfite
34449-10L-VP
34449-1L
BJ34449-10L-VPEA
641.45
USD
BJ34449-10L-VP
BJ-34449-1L
Sodium thiosulfate 0.05 M Na2S2O3 (0.05N) volumetric solution, Fluka™
Sodium thiosulphate
Formula:
Na₂S₂O₃ MW: 158.11 g/mol Boiling Pt: 100 Melting Pt: 48 Density: 1.0 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00003499 CAS Number: 7772-98-7 EINECS: 231-791-2 Merck Index: 12,08844 |
Specification Test Results
Factor | 0.999 - 1.001 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...