WesternBright ECL HRP Substrate, Advansta
Supplier: ADVANSTA INC MS
Advansta's HRP substrate is the best choice for routine, film-imaged chemiluminescent Western blots.
- Stronger signal than Amersham™ ECL or Pierce™ ECL
- Optimized for x-ray film imaging
- Sensitive for pg detection
- Lasting signal, up to 6 hrs
- Save money by using 10x less antibody
WesternBright ECL is a horseradish peroxidase substrate optimized for chemiluminescent Western blots imaged using X-ray film. WesternBright ECL is more sensitive than other chemiluminescent substrates and uses up to 10 times less antibody than other substrates, allowing you to save precious antibodies and samples. Additionally, the WesternBright ECL signal is long lasting, allowing multiple exposures without substantial signal decay.
Ordering information: Includes WesternBright ECL luminol/enhancer solution and WesternBright peroxide solution.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)