SHOWA Atlas 300 Natural Rubber Coated General-Purpose Work Glove, Showa
Supplier: Showa
300XXL-11 300M-08 300L-09
10847-722CS 282.53 USD
10847-722 10847-724 10847-784 10847-786 10847-788
SHOWA Atlas 300 Natural Rubber Coated General-Purpose Work Glove, Showa
Gloves
When handling extremely abrasive items and working in dirty environments, this glove provides longer wear and more protection than alternatives
  • Ergonomic design reduces hand fatigue
  • Durable rubber coating provides longer wear
Revered for comfortable contruction and tactical grip performance, this natural rubber palm coated, 10 gauge seamless knit glove is ergonomically designed to be the perfect fit for all general-purpose jobs.
Ability to launder extends use.
Ordering information: Order by Master case of 144 pair.
Packaging: Packaged 12 pair per poly bag, 12 bags per Master case.