QuantiFluor ONE dsDNA System, Promega
Supplier: Promega Corporation
The QuantiFluor(R) ONE dsDNA System enables sensitive quantitation of small amounts of double-stranded DNA (dsDNA). The system contains a fluorescent dsDNA-binding dye (504nm[Ex]/531nm[Em]) developed for use in an “add-and-read” format for dye and standard.
- Fluorescent Quantitation of dsDNA with "Add-and-Read" Convenience
- Specific quantitation of dsDNA
- Increased sensitivity over absorbance at 260nm for low-concentration samples
- Simple to use and automation-friendly
The QuantiFluor ONE dsDNA System contains a fluorescent double-stranded DNA-binding dye (504nmEx/531nmEm) developed for use in an “add-and-read” format for dye and standard, making sample quantitation easy. This system enables sensitive quantitation of small amounts of double-stranded DNA (dsDNA). The QuantiFluor ONE dsDNA System was developed using the fluorescence module of the GloMax Multi+ Detection System with Instinct Software, GloMax Discover System and the Quantus Fluorometer. The QuantiFluor ONE dsDNA System can be used with any fluorometer that is capable of measuring fluorescence at the appropriate excitation and emission wavelengths.
The QuantiFluor ONE dsDNA System can be used with any fluorometer that is capable of measuring fluorescence at the appropriate excitation and emission wavelengths. This system contains sufficient reagents for 100 or 500 assays at 200 μL scale.
The QuantiFluor ONE dsDNA System was developed using the fluorescence module of the GloMax® Multi+ Detection System with Instinct Software, GloMax Discover System, and the Quantus Fluorometer.