Soda lime for CO₂ absorption tube

Supplier: CE ELANTECH INC MS

338-352-35 338-352-30
101189-606EA 62.31 USD
101189-606 101189-604
Soda lime for CO₂ absorption tube
Soda lime
MDL Number: MFCD00134094
CAS Number: 8006-28-8
UN: 1907
ADR: 8 III
Merck Index: 13,08640

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR