SAF-T-FIT™ Plus P1130 Respirators, Honeywell Safety
Supplier: HONEYWELL SAFETY PRODUCTS USA INC.
SAF-T-FIT™
14110440 14110439
89013-638EA 9.7 USD
89013-638 89013-637
SAF-T-FIT™ Plus P1130 Respirators, Honeywell Safety
Respirators Disposable Respirators Particulate Respirators
SAF-T-FIT Plus molded cup disposable respirators offers variety in protection, size and comfort.
  • Nose clip is color-coded for visible size recognition
  • Latex free and durable suspension straps
  • Low breathing resistance
  • Humidity-resistant filter media