Purospher® STAR Phenyl HPLC Columns, MilliporeSigma
Supplier: MilliporeSigma
1.51920.0001 1.51921.0001 1.51922.0001 1.51920.1003 1.51921.1003 1.51922.1003
10143-998EA 892.08 USD
10143-998 10144-000 10144-002 10811-958 10811-960 10811-962
Purospher® STAR Phenyl HPLC Columns, MilliporeSigma
Chromatography Columns
Purospher® STAR Phenyl columns are based on high-purity silica particles, which provide symmetrical peaks for basic compounds, as well as high stability and excellent reproducibility.
  • Enhanced selectivity for aromatic compounds
  • Low silanol activity
  • pH stability from 1.0 to 10.5
  • Suitable for up to 100% aqueous mobile phases