Adhesive Vinyl GHS OSHA Labels 'Silica Amorphous', HCL Labels
Supplier: HCL Label
GH10604VN0023 GH10604VN0035 GH10604VN0047 GH20604VN0023 GH20604VN0035 GH20604VN0047 GH30604VN0023 GH30604VN0035 GH30604VN0047 GH40604VN0023 GH40604VN0035 GH40604VN0047
103023-070PK 74.03 USD
103023-070 103023-072 103023-074 103023-076 103023-078 103023-080 103031-112 103031-114 103031-116 103031-118 103031-120 103031-122 103039-000 103039-002 103039-004 103039-006 103039-008 103039-010 103046-604 103046-606 103046-608 103046-610 103046-612 103046-614
Adhesive Vinyl GHS OSHA Labels 'Silica Amorphous', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labelling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN Globally Harmonized System for the Classification and Labeling of Chemicals (GHS).
Certifications: RoHS compliant.