Adhesive Vinyl GHS OSHA Labels 'Hydrogen Cyanide', HCL Labels
Supplier: HCL Label
GH10519VN0023 GH10519VN0035 GH10519VN0047 GH20519VN0023 GH20519VN0035 GH20519VN0047 GH30519VN0023 GH30519VN0035 GH30519VN0047 GH40519VN0023 GH40519VN0035 GH40519VN0047
103022-494PK 74.03 USD
103022-494 103022-496 103022-498 103022-500 103022-502 103022-504 103030-104 103030-106 103030-108 103030-110 103030-112 103030-114 103038-288 103038-290 103038-292 103038-294 103038-296 103038-298 103045-708 103045-710 103045-712 103045-714 103045-716 103045-718
Adhesive Vinyl GHS OSHA Labels 'Hydrogen Cyanide', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN globally harmonized system for the classification and labeling of chemicals (GHS). All sizes are protected with a chemical resistant lamination and a shelf/outdoor life of two years.
Certifications: RoHS compliant