Adhesive Vinyl GHS OSHA Labels 'Aluminum', HCL Labels
Supplier: HCL Label
GH10415VN0023 GH10415VN0035 GH10415VN0047 GH20415VN0023 GH20415VN0035 GH20415VN0047 GH30415VN0023 GH30415VN0035 GH30415VN0047 GH40415VN0023 GH40415VN0035 GH40415VN0047
103020-922PK 74.03 USD
103020-922 103020-924 103020-926 103020-928 103020-930 103020-932 103028-730 103028-732 103028-734 103028-736 103028-738 103028-740 103036-580 103036-582 103036-584 103036-586 103036-588 103036-590 103044-648 103044-650 103044-652 103044-654 103044-656 103044-658
Adhesive Vinyl GHS OSHA Labels 'Aluminum', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN Globally Harmonized System for the Classification and Labeling of Chemicals (GHS).
Certifications: RoHS compliant.