BioClean™ Biotac™ Non sterile Nitrile Gloves, Ansell
Supplier: Ansell Healthcare
BIOTAC-L BIOTAC-M BIOTAC-S BIOTAC-XL BIOTAC-XS BIOTAC-XXL
75838-990CS 548.07 USD
75838-990 75838-992 75838-994 75838-996 75838-998 75839-000
BioClean™ Biotac™ Non sterile Nitrile Gloves, Ansell
Gloves
BioClean™ Biotac™ Non sterile Nitrile Gloves provide superior grip with excellent chemcial resistance.
  • Flexible and comfortable
  • Chemical resistant
  • Powder-free
  • Beaded cuff
  • Antistatic
  • Finger-textured for improved grip
  • ISO Class 5 compatible
  • Non-particulating EasyTear™ packaging
With superior grip and excellent ESD properties, BioClean™ Biotac™ Nitrile Gloves are latex-free, powder-free, and resistant to a wide range of chemicals. These ambidextrous, white, 30.5 cm (12") long nitrile cleanroom gloves feature beaded cuffs for strength and textured fingers for enhanced grip.
Certifications: Manufactured in a facility holding ISO 9001: 2008 and 89/686/EEC Article 11b certifications.
Conforms to Category 3 - Complex Design Personal Protective Equipment PPE Directive 89/686/EEC.
Processed in an NEBB certified ISO Class 5 environment.
Complies with the requirements of Glove Standard EN420:2003 and EN374-1, 2 & 3:2003.
Physical properties comply with European Medical Glove Standard EN455-2:2009.
Packaging: Gloves are packaged 100 pieces per sealed inner PE bag; one inner PE bag per sealed outer PE bag; 10 outer bags per lined carton (1000 pieces).