Whatman™ No. 1 Filter Paper, Whatman products (Cytiva)
Supplier: Cytiva
1001-042 1001-055 1001-070 1001-150 1001-185
470205-502PK 16.85 USD
470205-502 470205-504 470205-506 470205-508 470205-510
Whatman™ No. 1 Filter Paper, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
High Quality Filter Paper.
  • 11 µm particle retention
  • Particle retention rating at 98% efficiency
  • 180 µm nominal thickness
General purpose paper with medium flow and porosity. Ideal for gas filtration, air pollution analysis, and soil analysis.