3M™ 4520 Series Disposable Protective Coveralls, 3M Company
Supplier: 3M
7000088928 7000088929 7000088930 7000088931 7000088933 7000088934
89429-130CS 288.02 USD
89429-130 89429-132 89429-134 89429-136 89429-138 89429-140
3M™ 4520 Series Disposable Protective Coveralls, 3M Company
Disposable Garments
3M™ Disposable Protective Coverall Safety Work Wear 4520 helps protect against certain light liquid splashes and hazardous dusts. This coverall offers a collared option for situations where a hood is not required or may interfere with alternative head, face, or respiratory protection. Advanced, breathable and lightweight materials help reduce heat build-up and promote comfortable wear.
  • Protective coveralls protect against certain light liquid splashes and hazardous dusts
  • Lightweight, breathable material helps keeps workers cool and comfortable
  • Extra room in arms and legs allow for free movement on the job
  • Seamless shoulders and sleeve tops provide increased comfort and fewer potential entry points for contaminants
  • Meets government standards for Type 6 Splashes, Type 5 Dust, Anti-Static, and Nuclear protection
  • Garments do not contain components made from natural rubber latex to prevent reactions from users with latex sensitivities or restrictions.
3M™ Disposable Protective Coverall Safety Work Wear 4520 offers high-performance protection while delivering worker comfort
3M™ Protective Coveralls are designed for use in industrial workplaces as a physical barrier to help prevent skin contact from liquid splashes and dust in certain work environments. In addition, these coveralls meet ANSI 101-1996 size guidelines. Suggested applications include asbestos abatement and inspection, exposure to radioactive particles, lead abatement, mold remediation/mold spores, animal waste (solid), fertilizer application (dry/granular), pesticide application (dry/granular), dirt (general), general industrial clean-up and processing, grease (general), light-duty building cleaning, machine or vehicle maintenance, coal dust, electronics/ciruit board assembly, fiberglass/cellulose insulation installation, food processing, general powder handling, lime handling, warehousing, woodworking, non-hazardous aerosols (sprays), non-hazardous liquids, non-hazardous solids, metal particulate (chromium beryllium, etc.), and metal polishing.
In the "goods producing" sector of industry, which includes manufacturing, construction, and natural resources and mining, skin diseases/disorders rank as the second leading cause of incidence rates and nonfatal occupational illnesses in 2013 according to the Bureau of Labor Statistics. Protective apparel is a key factor in improving safety. These garments are only effective if they are worn consistently and matched to the hazards of the job.
These garments feature lightweight, breathable material technologies, extra room in the arms and legs for enhanced mobility, a two-way zipper for added on-off convenience, as well as storm flaps, seamless shoulders, and sleeve tops for extra protection. Coveralls are made from SMMMS material, which provides increased breathability and a barrier against hazardous particles and limited liquid splashes. All sizes have anti-static treatment on both sides, and are equipped with knit cuffs and elastic waist and ankles for increased comfort and mobility.
Certifications: Protective Clothing - standard EN 340, Type 6 Splash - EN 13034/(EN ISO 17491-4), Type 5 Dust - EN ISO 13982-1, Anti-Static* - EN 11492** and Nuclear - EN 1073 - 2.
Certified to European Standards to help protect against certain industrial hazards.
*Consult class results in Technical Data Sheet or User Instructions for more information.
**All apparel must be grounded for anti-static treatment to be effective.