Surgipath® Cut Resistant Gloves, Leica
Supplier: Leica Biosystems
3803550 3803555 3803560
75810-288PK 598.48 USD
75810-288 75810-290 75810-292
Surgipath® Cut Resistant Gloves, Leica
Gloves
Theese cut resistant gloves are manufactured from ultra-high molecular weight polyethylene, one of the strongest fibers made.
  • Lightweight and seamless
  • Very soft to touch
While handling sharp cutting instruments, the glove forms a barrier that helps protect the hand from cuts.
They can be worn between two latex, vinyl or nitrile gloves for maximum protection against body fluid contamination.These gloves can be worn time after time and sterilized between uses, however should be replaced when signs of wear appear.