Anti-CD90.2 Rat Monoclonal Antibody (PE) [clone: 30-H12]

Supplier: Tonbo Biosciences
50-0903-U025 50-0903-U100
75834-350EA 44 USD
75834-350 75834-352
Anti-CD90.2 Rat Monoclonal Antibody (PE) [clone: 30-H12]
Antibodies
The 30-H12 antibody reacts with mouse CD90.2 (Thy1.2). CD90.2 is a strain-specific allelic form of the GPI-linked membrane associated protein CD90 and is involved in adhesion and signal transduction. CD90.2 is expressed on thymocytes, mature T cells and neurons in mouse strains that express the CD90.2 allele (BALB/c, CBA, C3H, C57BL/6, SJL and others). 30-H12 does not react with the CD90.1 allele expressed in mouse strains such as PL and AKR.

Type: Primary
Antigen: CD90.2
Clonality: Monoclonal
Clone: 30-H12
Conjugation: PE
Epitope:
Host: Rat
Isotype: Rat IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR