Wave Apparatus
Supplier: VWR International
PH0712
470061-736EA 119.5 USD
470061-736
Wave Apparatus
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
A Classic Demonstration!
  • Demonstrate longitudinal and transverse motion with ease
This advanced apparatus demonstrates both longitudinal and transverse wave motions. Consisting of a number of eccentric discs supporting a series of metal rods revolving on a handle, transverse waves are obtained. Longitudinal waves are obtained from the bent rods. The rotating handle is equipped with a circular scale marked in degrees to indicate rotation. Includes instructions.