Thallium ICP Standard, Inorganic Ventures
Supplier: INORGANIC VENTURES MS
CGTL1-500ML CGTL10-500ML
100360-668EA 146.97 USD
100360-668 100360-174
Thallium ICP Standard, Inorganic Ventures
Standards Cationic Standards Thallium Standards
Certified reference material for Thallium, traceable to NIST.
One year expiration date (defined by supplier as the amount of time that it should remain in use after opening).