TransIT®-2020 Transfection Reagent, Mirus Bio
Supplier: Mirus Bio
TransIT®-2020 is a high-performance, animal-free broad spectrum DNA transfection reagent.
- Broad spectrum DNA delivery
- Gentle to cells
- Animal-free origin
TransIT®-2020 provides high performance with maximum compatibility so that you can achieve high expression in many cell types, including hard-to-transfect cell lines and primary cells.