Disposable Samplers, TargetDispo, Burkle
Supplier: BURKLE INC
5393-4460 5393-4461 5393-4470 5393-4471
10821-247PK 443.16 USD
10821-247 10821-249 10821-285 10821-287
Disposable Samplers, TargetDispo, Burkle
Samplers Disposable Samplers
TargetDispo is used to take a target point sample at a position in the sample material that has previously been precisely determined.
  • HDPE
  • Clean room manufactured
  • Optionally sterilised by gamma rays
The sampler pierces into the powder or granulate while closed. When it reaches the sample removal point it is opened and then closed again. In this way a precise target-point sample is obtained. The sample can simply be emptied through the open handle.