Methoxyacetonitrile ≥97%

Supplier: Matrix Scientific

114271-1G 114271-10G
102893-702EA 49.23 USD
102893-702 102893-704
Methoxyacetonitrile ≥97%
Methoxyacetonitrile

Hazard: Irritant

Formula: C₃H₅NO
MW: 71.08 g/mol
MDL Number: MFCD00001892
CAS Number: 1738-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR