3-(N-Boc-Methylaminomethyl)azetidine hydrochloride

Supplier: Matrix Scientific

Synonyms: 3-[(Methylamino)methyl]azetidine hydrochloride, 3-BOC protected

113416-1G 113416-5G
102892-134EA 727.69 USD
102892-134 102892-136
3-(N-Boc-Methylaminomethyl)azetidine hydrochloride
3-(N-Boc-Methylaminomethyl)azetidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₁ClN₂O₂
MDL Number: MFCD09998721
CAS Number: 1170019-97-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR