4',3,3-Trimethylbutyrophenone ≥97%

Supplier: Matrix Scientific

086740-1G 086740-2G 086740-5G
102850-342EA 672.31 USD
102850-342 102850-344 102850-346
4',3,3-Trimethylbutyrophenone ≥97%
4',3,3-Trimethylbutyrophenone

Hazard: Irritant

Formula: C₁₃H₁₈O
MDL Number: MFCD03841076
CAS Number: 681215-86-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR