2-(3-Methylbenzoyl)pyridine ≥97%
Supplier: Matrix Scientific
086599-1G
086599-2G
086599-5G
102849-508EA
795.38
USD
102849-508
102849-510
102849-512
2-(3-Methylbenzoyl)pyridine ≥97%
2-(3-Methylbenzoyl)pyridine
Hazard: Irritant
Formula:
C₁₃H₁₁NO |
MDL Number:
MFCD07699226 CAS Number: 59576-24-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...